Learn More
Synuclein α Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5038P
Description
This Anti-Synuclein Antibody, α is validated for use in immunohistochemistry (paraffin), western blotting for the detection of Synuclein.
Specifications
Synuclein α | |
Polyclonal | |
Affinity Purified immunoglobulin. Lyophilized, no preservatives. Reconstitute with 20μL of sterile distilled water. Centrifuge to remove any residue. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Rat |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
SNCA; PD1; alpha-synuclein; NACP; PARK4; MGC110988; PARK1; Alpha-synuclein | |
Human alpha-synuclein residues 111-131 coupled to KLH. The peptide sequence is N-GILEDMPVDPDNEAYEMPSEEC-C. This peptide is available for use in blocking studies (Cat. No. AG239). | |
20 μg | |
NM_000345.2; NM_007308.1 | |
−20-−70°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles. |