Learn More
Description
Specifications
Specifications
| Antigen | Synuclein α |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Lyophilized. Reconstitute with 50μL of sterile distilled water. Centrifuge to remove any residue. Glycerol (1:1) can be added for additional stability. |
| Gene Symbols | SNCA; PD1; alpha-synuclein; NACP; PARK4; MGC110988; PARK1; Alpha-synuclein |
| Host Species | Sheep |
| Immunogen | Synthetic peptide corresponding amino acids 116-131 of human alpha synuclein. |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.