Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Β-Synuclein Mouse anti-Human, Mouse, Rat, Clone: Syn207, MilliporeSigma™

Catalog No. 36009MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-009-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-009-MI Supplier MilliporeSigma Supplier No. 36009
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-β-Synuclein Antibody, clone Syn207 is an antibody against β-Synuclein for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen β-Synuclein
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone Syn207
Conjugate Unconjugated
Formulation ascites with 0.05% sodium azide
Gene Accession No. NM_001001502.1; NM_003085.3
Gene Symbols SNCB, beta-synuclein, Beta-synuclein.
Host Species Mouse
Immunogen full length recombinant human β-Synuclein
Purification Method Ascites
Quantity 200 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C for 24 months.
Form Ascites
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.