Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synuclein Β Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5086MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5086MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5086MI Supplier MilliporeSigma Supplier No. AB5086
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Synuclein Antibody, β is an antibody against Synuclein for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Synuclein β
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit serum. Lyophilized, no preservatives. Reconstitute with 50μL of sterile distilled water. Centrifuge to remove any residue.
Gene Symbols SNCB; beta-synuclein; Beta-synuclein.
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the carboxyl terminal of human beta-synuclein (IEPLMEPEGSYEDPPQE).
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_001001502.1; NM_003085.3
Target Species Human, Rat
Content And Storage Lyophilized: −20-−70°C for up to 6 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.