Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3, CD6 and CD20 cocktail Mouse, APC, FITC, PE, Clone: L27, 3G8, SP34-2, BD

Catalog No. BDB558639 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558639 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558639 Supplier BD Biosciences Supplier No. 558639
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cocktail Component Clone Isotype

PE anti-Human CD16 3G8 mIgG1

APC anti-Human CD20 L27 mIgG1

The NHP T/B/NK Cocktail is a three-color reagent cocktail designed to identify NHP T, B, and NK lymphocyte populations by direct immunofluorescent staining with flow cytometric analysis. Clone SP34-2 is a mouse IgG1 isotype monoclonal antibody, descendant of SP34 (mouse IgG3), with the same specificity and reactivity pattern as the parent clone. It cross-reacts with a major subset of peripheral blood lymphocytes, but not monocytes or granulocytes, of baboon, and rhesus, cynomolgus, and pigtail macaque monkeys. The monoclonal antibody clone L27 reacts with CD20 found only on B lymphocytes. The monoclonal antibody clone 3G8 reacts with CD16, the 50-65 kDa transmembrane form of IgG Fc (Fc γRIII), a human NK-cell-associated antigen. CD16 is expressed on NK cells as well as macrophages and granulocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3/CD6/CD20
Applications Flow Cytometry
Classification Monoclonal
Clone L27, 3G8, SP34-2
Conjugate FITC/APC/PE
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.