Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T-bet (TBX21) Mouse anti-Rat, Mouse, Human, Clone: 12E4.1, MilliporeSigma™

Catalog No. MABF80MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF80MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF80MI Supplier MilliporeSigma Supplier No. MABF80

Mouse Monoclonal Antibody

Detect T-bet (TBX21) using this Anti-T-bet (TBX21) Antibody, clone 12E4.1 validated for use in western blot, flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen T-bet (TBX21)
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 12E4.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_037483
Gene Symbols TBX21, TBET, TBLYM
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human TBX21.
Purification Method Purified by ion exchange chromatography
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.