Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
T-box transcription factor TBX5 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABE284
Description
Anti-T-box transcription factor TBX5 Antibody detects level of T-box transcription factor TBX5 and has been published and validated for use in Western Blotting, immunocytochemistry and immunohistochemistry.
Specifications
T-box transcription factor TBX5 | |
Polyclonal | |
Rabbit polyclonal serum with 0.05% sodium azide. | |
Rabbit | |
Unpurified | |
Primary | |
Human, Mouse, Rat | |
Serum |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Tbx5 | |
Recombinant protein corresponding to mouse T-box transcription factor TBX5. | |
100 μL | |
NP_035667 | |
-20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction