Learn More
Gammadelta T-Cell Receptor Hamster anti-Mouse, PE-CF594, Clone: GL3, BD
Hamster Monoclonal Antibody
Supplier: BD Biosciences 563532
Description
The GL3 monoclonal antibody specifically binds to a common epitope of the δ chain of the T-cell Receptor (TCR) complex on γδ TCR-expressing T lymphocytes and NK-T cells of all mouse strains tested. It does not react with αβ TCR-bearing T cells. In the mouse, cells expressing the γδ TCR are found in the thymus, intestinal epithelium, epidermis, dermis, pulmonsry epithelium, peritoneum, liver, and peripheral lymphoid organs.
Specifications
| Gammadelta T-Cell Receptor | |
| Monoclonal | |
| 0.2mg/mL | |
| Tcrd; T-cell receptor delta chain; Tcr delta | |
| Hamster | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2 κ |
| Flow Cytometry | |
| GL3 | |
| PE-CF594 | |
| Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
| C57BL/6 Mouse Intestinal Intraepithelial Lymphocytes | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.