Learn More
Description
Host Species: Hamster
Clone: GL3
Isotype: IgG2, κ
Species Reactivity: Ms
Immunogen: C57BL/6 Mouse Intestinal Intraepithelial Lymphocytes
Flow cytometry
Specifications
Specifications
| Antigen | gammadelta T-Cell Receptor |
| Classification | Monoclonal |
| Clone | GL3 |
| Concentration | 0.2mg/mL |
| Host Species | Hamster |
| Immunogen | C57BL/6 Mouse Intestinal Intraepithelial Lymphocytes |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.