Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

γδ T-Cell Receptor, Hamster anti-Mouse, Clone: GL3, BV711, BD

Catalog No. BDB563994 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563994 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563994 Supplier BD Biosciences Supplier No. 563994
Add to Cart
Add to Cart

Host Species: Hamster
Clone: GL3
Isotype: IgG2, κ
Species Reactivity: Ms
Immunogen: C57BL/6 Mouse Intestinal Intraepithelial Lymphocytes

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen gammadelta T-Cell Receptor
Classification Monoclonal
Clone GL3
Concentration 0.2mg/mL
Host Species Hamster
Immunogen C57BL/6 Mouse Intestinal Intraepithelial Lymphocytes
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.