Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

γδ T-Cell Receptor Mouse anti-Rat, PE, Clone: V65, BD

Catalog No. BDB551802 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551802 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551802 Supplier BD Biosciences Supplier No. 551802

Mouse Monoclonal Antibody

The V65 antibody reacts with the γδ T-cell Receptor (TCR) found on CD3+ αβ TCR-peripheral T lymphocytes, on a small subset of intestinal intraepithelial lymphocytes, and on dendritic epidermal T cells. Cross-linked V65 mAb induces cytokine-dependent T-cell mitogenesis in vitro , and in vivo treatment can significantly deplete peripheral γδ TCR-expressing T cells. A suspension of rat CD3+ αβ TCR-T-cell hybridoma III.89.1.4 was used as the source of the immunogen.

TRUSTED_SUSTAINABILITY

Specifications

Antigen γδ T-Cell Receptor
Applications Flow Cytometry
Classification Monoclonal
Clone V65
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat CD3+ αβ TCR- T-cell hybridoma III.89.1.4
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.