Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T, brachyury homolog (mouse), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018301 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-301 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-301 Supplier Abnova Corporation Supplier No. H00006862D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human T protein.

The protein encoded by this gene is an embryonic nuclear transcription factor that binds to a specific DNA element, the palindromic T-site. It binds through a region in its N-terminus, called the T-box, and effects transcription of genes required for mesoderm formation and differentiation. The protein is localized to notochord-derived cells. [provided by RefSeq

Sequence: MSSPGTESAGKSLQYRVDHLLSAVENELQAGSEKGDPTERELRVGLEESELWLRFKELTNEMIVTKNGRRMFPVLKVNVSGLDPNAMYSFLLDFVAADNHRWKYVNGEWVPGGKPEPQAPSCVYIHPDSPNFGAHWMKAPVSFSKVKLTNKLNGGGQIMLNSLHKYEPRIHIVRVGDPQRMITSHCFPETQFIAVTAYQNEEITALKIKYNPFAKAFLDAKERSDHKEMMEEPGDSQQPGYSQSYSDNSPACLSMLQSHDNWSSLGMPAHPSMLPVSHNASPPTSSSQYPSLWSVSNGAVTPGSQAAAVSNGLGAQFFRGSPAHYTPLTHPVSAPSSSGSPLYEGAAAATDIVDSQYDAAAQGRLIASWTPVSPPSM

Specifications

Antigen T, brachyury homolog (mouse)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human T protein.
Formulation PBS with no preservative; pH 7.4
Gene T
Gene Accession No. BC098425.1
Gene Alias MGC104817/TFT
Gene Symbols T
Host Species Rabbit
Immunogen T (AAH98425.1, 1 a.a. ∼ 377 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Gene ID (Entrez) 6862
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.