Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TM4SF4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526753
Description
T4S4 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that can regulate cell proliferation. The use of alternate polyadenylation sites has been found for this gene.
Specifications
| TM4SF4 | |
| Polyclonal | |
| TM4SF4 | |
| ILTMP, il-TMP, intestinal and liver (il) tetraspan membrane protein, intestine and liver tetraspan membrane protein, transmembrane 4 L6 family member 4, transmembrane 4 superfamily member 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 229302, 7104 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| P48230, Q91XD3 | |
| TM4SF4 | |
| KLH conjugated synthetic peptide between 16-45 amino acids from the N-terminal region of human T4S4 | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction