Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TC2N Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526433
Description

Specifications
TC2N | |
Polyclonal | |
TC2N | |
C14orf47, C2CD1, MTAC2D1, Tac2-N, C2 calcium-dependent domain containing 1, membrane targeting (tandem) C2 domain containing 1, membrane targeting tandem C2 domain-containing protein 1, tandem C2 domains nuclear protein, tandem C2 protein in nucleus | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
123036, 74413 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q8N9U0, Q91XT6 | |
TC2N | |
KLH conjugated synthetic peptide between 3-30 amino acids from the N-terminal region of human TAC2N | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction