Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-TACE (807 - 823), Polyclonal

Catalog No. PC491100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
PC491100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PC491100UG Supplier MilliporeSigma™ Supplier No. PC491100UG

Rabbit Polyclonal Antibody

Specifically detects TACE (807 - 823) Clone: in Human, Mouse, Rat samples, and it is validated for Immuno Blotting

Tumor-necrosis factor-α is a proinflammatory cytokine that contributes to a variety of inflammatory disease responses and programmed cell death. TNF-α is synthesized as a 26K type II membrane-bound precursor that is cleaved by a convertase to generate secreted 17K mature TNF-α. TNF-α converting enzyme (TACE) protein was recently purified and the human and mouse TACE cDNAs were cloned by several groups separately. TACE, a∽130kDa protein, is a membrane-bound metalloprotease-disintegrin in the family of mammalian ADAM (for disintegrin and metalloprotease). TACE also processes other cell surface proteins, including TNF receptor, TGFα, the L-selectin adhesion molecule, and alpha-cleavage of amyloid protein precursor (APP). TACE mRNA is expressed in a variety of human and murine tissues. TACE was selected as one of the few targets in cytokine activation by the Eighth International Conference of the Inflammation Research Association.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TACE (807 - 823)
Applications Immunoblot
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS.
Host Species Rabbit
Immunogen a synthetic peptide (ASFKLQRQNRVDSKETE) corresponding to amino acids 807-823 of human TACE
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽130kDa TACE protein in HeLa and Jurkat cells.
Target Species Human, Mouse, Rat
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.