Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TACI Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABC847
Description
Detect TACI using this Anti-TACI Antibody validated for use in western blotting and immunocytochemistry.
Specifications
TACI | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
2-8°C for 12 months |
Immunocytochemistry, Western Blot | |
Unconjugated | |
TNFRSF13B; TACI | |
KLH-conjugated synthetic peptide corresponding to the N-terminus of human TACI. | |
100 μg | |
Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction