Learn More
TAF1A, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA124350
Description
Initiation of transcription by RNA polymerase I requires the formation of a complex composed of the TATA-binding protein (TBP) and three TBP-associated factors (TAFs) specific for RNA polymerase I. This complex, also known as SL1, binds to the core promoter of ribosomal RNA genes to position the polymerase properly and acts as a channel for regulatory signals. This gene encodes the smallest SL1-specific TAF. Two transcripts encoding different isoforms have been identified.
PA1-24350 detects TAF(I)48 in Human samples.PA1-24350 has been successfully used in Western Blot procedures.PA1-24350 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human TAF1A.
Specifications
TAF(I)48 | |
Polyclonal | |
Unconjugated | |
Q15573 | |
Rabbit | |
Protein A/G | |
RUO | |
9015 | |
Store at -20°C, Avoid Freeze/Thaw Cycles | |
Purified |
Western Blot | |
1mg/mL | |
TAF1A | |
TAF1A | |
Synthetic peptide region of human TAF1A. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.