Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF(II)250 Rabbit anti-Human, Mouse, Rat, Bovine, Horse, Canine, Polyclonal, MilliporeSigma™

Catalog No. ABE42MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE42MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE42MI Supplier MilliporeSigma Supplier No. ABE42

Rabbit Polyclonal Antibody

Use Anti-TAF(II)250 Antibody (rabbit polyclonal antibody) validated in western blotting to detect TAF(II)250 also known as Transcription initiation factor TFIID subunit 1, Cell cycle gene 1 protein, TBP-associated factor 250 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF(II)250
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols TAF1; BA2R; CCG1; CCGS; TAF2A
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human TAF(II)250 at the protein kinase 2 domain.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_004597
Target Species Bovine, Canine, Equine, Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.