Learn More
TAF11 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 28kDa, Mouse, Clone: 3D3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant TAF11.
Supplier: Abnova Corporation H00006882M01
Description
Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes a small subunit of TFIID that is present in all TFIID complexes and interacts with TBP. This subunit also interacts with another small subunit, TAF13, to form a heterodimer with a structure similar to the histone core structure. [provided by RefSeq
Sequence: SKVFVGEVVEEALDVCEKWGEMPPLQPKHMREAVRRLKSKGQIPNSKHKKIIFSpecifications
TAF11 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 28kDa | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_005643 | |
TAF11 | |
TAF11 (NP_005634, 158 a.a. ∼ 210 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Transcription Regulation | |
Primary | |
Human | |
Antibody |
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
3D3 | |
Mouse monoclonal antibody raised against a partial recombinant TAF11. | |
TAF11 | |
MGC:15243/PRO2134/TAF2I/TAFII28 | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
6882 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.