Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF15 Mouse anti-Human, Mouse, Rat, Clone: 8TA-2B10, MilliporeSigma™

Catalog No. MABE450MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE450MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE450MI Supplier MilliporeSigma Supplier No. MABE450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-TAF15 antibody, clone 8TA-2B10 is an antibody against TAF15 for use in western blotting and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF15
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 8TA-2B10
Conjugate Unconjugated
Formulation Mouse monoclonal IgG1κ ascites without preservatives.
Gene Accession No. NP_003478
Gene Symbols TAF15, RBP56, TAF2N
Host Species Mouse
Immunogen Recombinant protein corresponding to human TAF15.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.