Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kDa, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016960 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-960 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-960 Supplier Abnova Corporation Supplier No. H00009013B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human TAF1C protein.

Initiation of transcription by RNA polymerase I requires the formation of a complex composed of the TATA-binding protein (TBP) and three TBP-associated factors (TAFs) specific for RNA polymerase I. This complex, known as SL1, binds to the core promoter of ribosomal RNA genes to position the polymerase properly and acts as a channel for regulatory signals. This gene encodes the largest SL1-specific TAF. Two transcripts encoding different isoforms have been identified. [provided by RefSeq

Sequence: MLPPLIDPWDPGLTARDLLFRGGYRYRKRPRVVLDVTEQISRFLLDHGDVAFAPLGKLMLENFKLEGAGSRTKKKTVVSVKKLLQDLGGHQPWGCPWAYLSNRQRRFSILGGPILGTSVASHLAELLHEELVLRWEQLLLDEACTGGALAWVPGRTPQFGQLVYPAGGAQDRLHFQEVVLTPGDNPQFLGKPGRIQLQGPVRQVVTCTVQGETLLAVRSDYHCAVWKFGKQWQPTLLQAMQVEKGATGISLSPHLPGELAICSRSGAVCLWSPEDGLRQIYRDPETLVFRDSSSWRWADFTAHPRVLTVGDRTGVKMLDTQGPPGCGLLLFRLGAEASCQKGERVLLTQYLGHSSPKCLPPTLHLVCTQFSLYLVDERLPLVPMLKWNHGLPSPLLLARLLPPPRPSCVQPLLLGGQGGQLQLLHLAEGASVPRLAGPPQSLPSRIDSLPAFPLLEPKIQWRLQERLKAPTIGLAAVVPPMPSAPTPGLVLFQLSAAGDVFYQQLRPQVDSSLRRDAGPPGDTQPDCHAPTASWTSQDTAGCSQWLKALLKVPLAPPVWTAPTFTHRQMLGSTELRREEEEGQRLGVLRKAMARGQLLLQRDLGSLPAAEPPPAPESGLEDKLSERLGEAWAGRGAAWWERQQGRTSEPGRQTRRPKRRTQLSSSFSLSGHVDPSEDTSSPHSPEWPPADALPLPPTTPPSQELTPDACAQGVPSEQRQMLRDYMAKLPPQRDTPGCATTPPHSQASSVRATRSQQHTPVLSSSQPLRKKPRMGF

Specifications

Antigen TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kDa
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human TAF1C protein.
Formulation PBS with no preservative; pH 7.4
Gene TAF1C
Gene Accession No. BC028131.1
Gene Alias MGC:39976/SL1/TAFI110/TAFI95
Gene Symbols TAF1C
Host Species Mouse
Immunogen TAF1C (AAH28131.1, 1 a.a. ∼ 775 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Gene ID (Entrez) 9013
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.