Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF3 Rabbit anti-Mouse, Rhesus Macaque, Guinea Pig, Polyclonal, MilliporeSigma™

Catalog No. 071802MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-180-2MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-180-2MI Supplier MilliporeSigma Supplier No. 071802
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect TAF3 using this Anti-TAF3 Antibody validated for use in Chromatin Immunoprecipitation (ChIP) and Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF3
Applications ChIP Assay, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum containing with 0.05% sodium azide.
Gene Symbols TAF3
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to human TAF3.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_114129.1
Target Species Guinea Pig, Mouse, Rhesus Monkey
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.