Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 55kDa, Mouse, Clone: 3G6, Abnova™

Catalog No. 89000997 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-997 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-997 Supplier Abnova Corporation Supplier No. H00006879M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant TAF7.

The intronless gene for this transcription coactivator is located between the protocadherin beta and gamma gene clusters on chromosome 5. The protein encoded by this gene is a component of the TFIID protein complex, a complex which binds to the TATA box in class II promoters and recruits RNA polymerase II and other factors. This particular subunit interacts with the largest TFIID subunit, as well as multiple transcription activators. The protein is required for transcription by promoters targeted by RNA polymerase II. [provided by RefSeq

Sequence: FIWNHGITLPLKNVRKRRFRKTAKKKYIESPDVEKEVKRLLSTDAEAVSTRWEIIAEDETKEAENQGLDISSPGMSGHRQGHDSLEHDELREIFN

Specifications

Antigen TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 55kDa
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3G6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant TAF7.
Formulation PBS with no preservative; pH 7.4
Gene TAF7
Gene Accession No. BC032737
Gene Alias TAF2F/TAFII55
Gene Symbols TAF7
Host Species Mouse
Immunogen TAF7 (AAH32737, 130 a.a. ∼ 224 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 6879
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.