Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Talin 1 Mouse anti-Human, Rabbit, Clone: TA205, MilliporeSigma™

Catalog No. MAB1676MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1676MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1676MI Supplier MilliporeSigma Supplier No. MAB1676
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Talin 1 Antibody, N-terminus, a.a. 139-433, clone TA205 detects level of Talin and has been published and validated for use in immunoprecipitation, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Talin 1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone TA205
Conjugate Unconjugated
Formulation Liquid in 0.02MPB pH 7.6, 0.25MNaCl containing 0.1% sodium azide.
Gene Accession No. NM_006289.2
Gene Symbols TLN1, talin, ILWEQ, KIAA1027, TLN, Talin-1.
Host Species Mouse
Immunogen Human platelet protein
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Rabbit
Content And Storage 2-8°C.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.