Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAP Mouse, Unlabeled, Clone: 31, BD

Catalog No. BDB611586 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611586 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611586 Supplier BD Biosciences Supplier No. 611586

Mouse Monoclonal Antibody

Nuclear export of cellular mRNAs is highly selective, wherein only fully processed RNAs are exported. Retroviral replication requires export of unspliced viral RNAs, which serve as templates for structural proteins and as genomic RNA for progeny virus. The export of unspliced viral RNA depends on the interaction of cis-acting RNA elements with cellular factors. In simian type D retroviruses, the cis-acting element is the constitutive transport element (CTE). CTE-dependent nuclear transport is mediated by the cellular TAP protein. TAP (Tip associating protein) is the vertebrate homolog of the yeast nuclear export protein Mex67p. TAP is a nuclear protein that shuttles between the nucleus and cytoplasm. It contains an N-terminal nuclear localization and export region (NLS-NES), a central RNA binding domain (RBD), and a C-terminal portion that is required for nuclear localization and nuclear rim association. The first 372 aa of TAP are necessary and sufficient for binding and nuclear export of CTE-containing RNA. Thus, TAP contains a novel RNA-binding motif that recognizes CTE-containing RNA and interacts with other components of the nuclear transport machinery.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TAP
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 31
Conjugate Unconjugated
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Rat
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.