Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAP Mouse, Unlabeled, Clone: 19, BD

Catalog No. BDB611580 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611580 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611580 Supplier BD Biosciences Supplier No. 611580
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nuclear export of cellular mRNAs is highly selective, wherein only fully processed RNAs are exported. Retroviral replication requires export of unspliced viral RNAs, which serve as templates for structural proteins and as genomic RNA for progeny virus. The export of unspliced viral RNA depends on the interaction of cis-acting RNA elements with cellular factors. In simian type D retroviruses, the cis-acting element is the constitutive transport element (CTE). CTE-dependent nuclear transport is mediated by the cellular TAP protein. TAP (Tip associating protein) is the vertebrate homolog of the yeast nuclear export protein Mex67p. TAP is a nuclear protein that shuttles between the nucleus and cytoplasm. It contains an N-terminal nuclear localization and export region (NLS-NES), a central RNA binding domain (RBD), and a C-terminal portion that is required for nuclear localization and nuclear rim association. The first 372 aa of TAP are necessary and sufficient for binding and nuclear export of CTE-containing RNA. Thus, TAP contains a novel RNA-binding motif that recognizes CTE-containing RNA and interacts with other components of the nuclear transport machinery.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TAP
Applications Western Blot
Classification Monoclonal
Clone 19
Concentration 250μg/mL
Conjugate Unconjugated
Description Manganese Superoxide Dismutase
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human TAP aa. 488-596
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.