Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tapasin Rat anti-Human, Clone: 7F6, MilliporeSigma™

Catalog No. MABF249MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF249MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF249MI Supplier MilliporeSigma Supplier No. MABF249
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Detect Tapasin using this rat monoclonal antibody, Anti-Tapasin Antibody, clone 7F6 validated for use in western blotting and Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tapasin
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 7F6
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. np_003181
Gene Symbols TAPBP, NGS17, TAPA
Host Species Rat
Immunogen Linear peptide corresponding to human Tapasin.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.