Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-TARDBP Clone: 2E2-D3,

Catalog No. DR1075100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
DR1075100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DR1075100UG Supplier MilliporeSigma™ Supplier No. DR1075100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects TARDBP Clone: 2E2-D3 in Human samples, and it is validated for Immunocytochemistry, Immunohistochemistry (Paraffin)

HIV-1, the causative agent of acquired immunodeficiency syndrome (AIDS), contains an RNA genome that produces a chromosomally integrated DNA during the replicative cycle. Activation of HIV-1 gene expression by the transactivator Tat is dependent on an RNA regulatory element (TAR) located downstream of the transcription initiation site. TARDBP is a transcriptional repressor that binds to chromosomally integrated TAR DNA and represses HIV-1 transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TARDBP
Applications Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2E2-D3
Conjugate Unconjugated
Formulation In PBS, pH 7.7.
Host Species Mouse
Immunogen Full-length, recombinant human TARDBP (aa 1-261), expressed as a GST fusion protein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽45kDa TARDBP protein in A431 and HeLa cells and human leiomyosarcoma tissue.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.