Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAT, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA127504
Description
Transcriptional regulator that acts by binding to the trans-activating responsive sequence (TAR) RNA element and activates transcription initiation and/or elongation from the LTR promoter.
PA1-27504 detects tat from human, mouse, and rat samples.PA1-27504 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27543 immunogen is fragment of Protein Tat.
Specifications
tat | |
Polyclonal | |
Unconjugated | |
P17735, Q8QZR1, P04694 | |
Chicken | |
Antigen affinity chromatography | |
RUO | |
6898 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
Western Blot | |
1mg/mL | |
TAT | |
TAT | |
Fragment of Protein Tat. | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction