Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tau (3-repeat isoform RD3) Mouse anti-Human, Clone: 8E6/C11, MilliporeSigma™

Catalog No. 05803MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-803-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-803-MI Supplier MilliporeSigma Supplier No. 05803
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Tau (3-repeat isoform RD3) Antibody, clone 8E6/C11 is an antibody against Tau (3-repeat isoform RD3) for use in immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tau (3-repeat isoform RD3)
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 8E6/C11
Conjugate Unconjugated
Formulation Mouse culture supernatant containing 0.05% sodium azide. Frozen at −20°C.
Gene Accession No. NM_016835
Gene Symbols MAPT, MTBT2, tau, FTDP-17, MSTD, TAU, MTBT1, PHF-tau, MGC138549, MAPTL, FLJ31424, DDPAC, PPND
Host Species Mouse
Immunogen Bovine thyroglobulin conjugated synthetic peptide corresponding to amino acids 209-224 (KHQPGGGKVQIVYKPV) of human Tau (isoform RD3). In other isoforms of huTau this sequence spans amino acids 267-316, omitting the second repeat domain where it bridges RD
Purification Method Unpurified
Quantity 200 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.