Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Tau (4-repeat isoform RD4), Clone: 1E1/A6, ™ (Upstate™)
Supplier: MilliporeSigma™ 05804/DEL
Description
Utton, M.A. et al. J. Neurosci., 22: 6394-6400, 2002.
Immunohistochemistry, Western Blotting
Specifications
| Tau (4-repeat isoform RD4) | |
| Unconjugated | |
| Bovine thyroglobulin-conjugated synthetic peptide corresponding to amino acids 275-291 (VQIINKKLDLSNVQSKC) of human Tau (4-repeat isoform RD4). Region is flanking junction coded by adjacent exons 9 & 11 with the inclusion of exon 10. | |
| RUO | |
| Primary | |
| Supernatant |
| Monoclonal | |
| Mouse | |
| 200 μL | |
| Neuroscience | |
| Bovine, Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction