Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tau Mouse anti-Bovine, Human, Mouse, Rat, Rabbit, Clone: 5E2, MilliporeSigma™

Catalog No. 05348MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-348-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-348-MI Supplier MilliporeSigma Supplier No. 05348
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Tau using this Anti-Tau Antibody, clone 5E2 validated for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tau
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 5E2.
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, containing and 0.05% sodium azide
Gene Accession No. NM_016835
Gene Symbols MAPT, MTBT2, tau, FTDP-17, MSTD, TAU, MTBT1, PHF-tau, MGC138549, MAPTL, FLJ31424, DDPAC, PPND
Host Species Mouse
Immunogen Fetal heat stable MAPS
Purification Method Protein G chromatography
Quantity 200 μg
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Rabbit, Rat
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.