Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tau Mouse anti-Human, Clone: Tau 12, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB2241
Description
Anti-Tau Antibody, clone Tau 12 is an antibody against Tau for use in western blotting.
Specifications
Tau | |
Monoclonal | |
Unconjugated | |
P10636 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Western Blot | |
Tau 12 | |
Purified in 0.1M Tris-Glycine (pH7.4) 150mM NaCl with 0.05% NaN3. | |
DDPAC , FLJ31424, FTDP-17 , MAPTL, MSTD , MTBT1, MTBT2 , PHF-tau , PPND , TAU, tau | |
KLH conjugated synthetic linear peptide. | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction