Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Tau Clone: Tau-2,

Catalog No. MAB375CMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MAB375CMI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB375CMI Supplier MilliporeSigma™ Supplier No. MAB375C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Tau Clone: Tau-2 in Human, Rat samples, and it is validated for Immunohistochemistry

Microtubule-associated protein tau (UniProt P29172; also known as Neurofibrillary tangle protein, Paired helical filament-tau, PHF-tau) is encoded by the MAPT (also known as TAU) gene (Gene ID 281296) in bovine species. Extracellular plaque deposits composed of amyloid-β and intracellular neurofibrillary tangles (NFTs) composed of truncated and hyperphosphorylated tau are two neuropathological hallmarks of Alzheimer's disease (AD). Tau pathology (tauopathy) can cause toxicity when the brain is devoid of amyloid plaques, and tangle pathology correlates better with clinical dementia than amyloid pathology. Abnormal processing of tau by hyperphosphorylation and proteolytic truncation contribute to the toxicity of tau. Tau is known to be cleaved by various proteases, including caspases, calpains, cathepsins, thrombin, and puromycin-sensitive aminopeptidase, as well as the lysosomal cysteine proteinase asparagine endopeptidase (AEP). MAPK, GSK-3, and Cdk-5 are three known kinases that target various tau phosphorylation sites. Tau also undergoes other types of posttranslational modifications, including glycosylation, ubiquitination, glycation, polyamination, nitration, and lysine methylation, which are believed to be important for its non-pathological functions, including polymerization and stabilization of microtubules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tau
Applications Immunohistochemistry
Classification Monoclonal
Clone Tau-2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P29172
Gene Symbols MAPT, TAU
Host Species Mouse
Immunogen Bovine Tau.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_776531
Test Specificity Tau from bovine, feline and abnormal human tau in Alzheimer's disease.
Target Species Human, Rat
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.