Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Tau Clone: TAU-5

Catalog No. 577801100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
57-780-1100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57-780-1100UG Supplier MilliporeSigma™ Supplier No. 577801100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Tau Clone: TAU-5 in Human, Mouse, Rat, Sheep samples, and it is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Immuno Blotting

Tau is a neuronal microtubule-associated protein found predominantly in axons. Tau promotes tubulin polymerization and stabilizes microtubules, and it links certain signaling pathways to the cytoskeleton. Hyperphosphorylated tau is the major component of paired helical filaments, the building block of neurofibrillary lesions in Alzheimer’s disease. Hyperphosphorylation impairs the microtubule-binding function of tau, resulting in the destabilization of microtubules, and ultimately leading to degeneration of the affected neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tau
Applications Immunoblot, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone TAU-5
Conjugate Unconjugated
Formulation In PBS.
Host Species Mouse
Immunogen purified bovine microtubule-associated proteins
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽45-68kDa Tau protein.
Target Species Human, Mouse, Rat, Sheep
Content And Storage ≤ -70°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.