Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tau Mouse anti-Bovine, Human, Mouse, Rat, Clone: Tau-5, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB361
Description
Anti-Tau Antibody, a.a. 210-241, clone Tau-5 detects level of Tau and has been published and validated for use in immunohistochemistry and western blotting.
Specifications
Tau | |
Monoclonal | |
Unconjugated | |
NM_005910.3 | |
Mouse | |
Unpurified | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
Immunohistochemistry, Western Blot | |
Tau-5 | |
Unpurified mouse monoclonal IgG1 ascites fluid containing no preservatives. | |
MAPT, MTBT2, MAPTL, PHF-tau, tau, DDPAC, FTDP-17, MGC138549, MSTD, TAU, FLJ31424, MTBT1, PPND | |
Bovine tau | |
100 μL | |
Bovine, Human, Mouse, Rat | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction