Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tau Mouse anti-Human, Rat, Clone: Tau 7, MilliporeSigma™

Catalog No. MAB2239MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2239MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2239MI Supplier MilliporeSigma Supplier No. MAB2239
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Tau Antibody, clone Tau 7 is validated for use in western blotting for the detection of Tau.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tau
Applications Western Blot
Classification Monoclonal
Clone Tau 7
Conjugate Unconjugated
Formulation Purified in 0.1M Tris-Glycine (pH7.4) 150mM NaCl with 0.05% NaN3.
Gene Accession No. NP_001116538.1
Gene Symbols DDPAC, FLJ31424, FTDP-17, MAPTL, MGC138549 , MSTD, MTBT1, MTBT2, PHF-tau, PPND, TAU, tau
Host Species Mouse
Immunogen KLH conjugated synthetic linear peptide.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.