Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tau Mouse anti-Human, Mouse, Rat, Clone: TNT-1, MilliporeSigma™

Catalog No. MABN471MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN471MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN471MI Supplier MilliporeSigma Supplier No. MABN471
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Tau, PAD, clone TNT-1 detects levels of Tau proteins and has been published and validated for use in western blot, immunohistochemistry, immunofluorescence and ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tau
Applications ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone TNT-1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001116538
Gene Symbols MAPT, MAPTL, MTBT1, TAU
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to human Tau.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.