Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tau (T22), oligomeric Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABN454MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN454MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN454MI Supplier MilliporeSigma Supplier No. ABN454
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Tau (T22) antibody, oligomeric is validated for use in Dot Blot, ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Neutralization, and Western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tau (T22)
Applications Dot Blot, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Neutralization, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum containing 0.05% sodium azide.
Gene Symbols MAPT; MAPTL; MTBT1; TAU
Host Species Rabbit
Immunogen Tau oligomers corresponding to human oligomeric Tau (T22).
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_001116538
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.