Learn More
Taurine Transporter Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Taurine Transporter |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in PBS containing 0.1% BSA and 0.05% sodium azide. |
| Gene Symbols | SC6A6; SLC6A6 |
| Host Species | Rabbit |
| Immunogen | A 20 amino acid peptide near the carboxy terminal cytoplasmic region of rat Taurine Transporter (Smith et al. 1992; Liu et al. 1993; Uchida et al. 1992; Jhiang et al. 1993). |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.