Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Taurine Transporter Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB2235MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2235MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2235MI Supplier MilliporeSigma Supplier No. AB2235

Rabbit Polyclonal Antibody

This Anti-Taurine Transporter Antibody is validated for use in western blotting for the detection of Taurine Transporter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Taurine Transporter
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified in 0.1M Tris-Glycine (pH7.4) 15mM NaCl with 0.05% NaN3
Gene Symbols MGC10619; MGC131729; TAUT 1
Host Species Rabbit
Immunogen Synthetic Linear Peptide.
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001134367.1
Target Species Rat
Content And Storage 2-8°C for 12 months in undiluted aliquotes
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.