Learn More
TBC1D7, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human TBC1D7 peptide using ARM Technology.
Supplier: Abnova Corporation H00051256K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
TBC1D7 belongs to the family of proteins sharing a 180- to 200-amino acid TBC domain presumed to have a role in regulating cell growth and differentiation. These proteins share significant homology with TRE2 (USP6; MIM 604334), yeast Bub2, and CDC16 (MIM 603461) (Nakashima et al., 2007 [PubMed 17658474]).[supplied by OMIM
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
| TBC1D7 | |
| Monoclonal | |
| Unconjugated | |
| In 1X PBS, pH 7.4 | |
| TBC1D7 | |
| TBC1D7 | |
| A synthetic peptide of human TBC1D7 | |
| RUO | |
| 51256 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| ELISA, Western Blot | |
| 1mg | |
| TBC1 domain family, member 7 | |
| TBC1D7 | |
| DKFZp686N2317, FLJ32666, PIG51, TBC7, dJ257A7.3 | |
| Rabbit | |
| 1 mg x up to 3 Abs | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.