Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBK1 Mouse anti-Human, Alexa Fluor 647, Clone: 637IG11.2, BD

Catalog No. BDB558698 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558698 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558698 Supplier BD Biosciences Supplier No. 558698

Mouse Monoclonal Antibody

NF-κB is a ubiquitously expressed transcription factor that regulates many cytokine and Ig genes. It is involved in immune, inflammatory, viral, and acute phase responses. In most cells, NF-κB is sequestered in an inactive cytoplasmic form via interactions with the inhibitory proteins IκBα, IκBβ, and IκBε. Stimulation induces the release, activation, and nuclear translocation of NF-κB. Release of NF-κB results from the phosphorylation and proteolytic degradation of the IκB proteins. Two cytokine-inducible IκB kinases (IKKα and IKKβ) phosphorylate and target the IκB proteins for degradation via the ubiquitin pathway. IKKγ/NEMO, a third member of the IKK complex, functions as a regulatory subunit and interacts directly with IKKβ. TBK1 (TANK-binding kinase 1, also known as T2K or NAK), a protein of 729 amino acids, is another member of the IKK family of kinases regulating NF-κB downstream of the tumor necrosis factor and Toll-like receptor pathways. TBK1 forms a complex with the adaptor proteins TANK (TRAF-associated NF-κB activator) and TRAF2 (TNF-receptor-associated factor 2), and this oligomer is required for activation and phosphorylation of TBK1 at serine 172. The 637Ig11.2 monoclonal antibody recognizes human TBK1, regardless of phosphorylation status. Our in-house testing is performed on a cell line that has been co-transfected with TBK1 and IRF-7 because we have been unable to detect endogenous TBK1 by western blotting of lysates from activated cell lines (HeLa activated with PMA and A431 activated with PDGF). In the transfectants, the over-expression of TBK1 is necessary for phosphorylation of IRF-7, but the presence of IRF-7 does not affect TBK1 expression or phosphorylation. We confirmed that mAb 637Ig11.2 does not cross-react with IRF-7 by western blot analysis using the purified antibody.

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen TBK1
Applications Flow Cytometry
Classification Monoclonal
Clone 637IG11.2
Conjugate Alexa Fluor 647
Description T2K, NAK
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human TBK1 C terminus Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.