Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-TBK1 Clone: 108A429,

Catalog No. AM76100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
AM76100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM76100UG Supplier MilliporeSigma™ Supplier No. AM76100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects TBK1 Clone: 108A429 in Human samples, and it is validated for Immuno Blotting

Nuclear factor κB (NF-κB) is sequestered in cell cytoplasm by the IκB family of inhibitory proteins that mask the nuclear localization signal of NF-κB, preventing its translocation to the nucleus. Stimulus by tumor necrosis factor or other cytokines results in the phosphorylation and degradation of IκB, releasing NF-κB dimers. These dimers then translocate to the nucleus and activate target genes. IκBα synthesis is autoregulated. IκB proteins are phosphorylated by an IκB kinase made up of at least three proteins; IKK1/α, IKK2/β, and IKK3/γ. IKK 1/α and IKK2/β are phosphorylated by NF-κB-inducing kinase (NIK) and MAP kinase kinase kinase-1 (MEKK1) respectively. TANK-binding kinase (TBK1) has recently been identified as a member of the IKK family. TRAF-associated NF-κB activator (TANK) interacts with TBK1 and activates NF-κB in a kinase-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBK1
Applications Immunoblot
Classification Monoclonal
Clone 108A429
Conjugate Unconjugated
Formulation In PBS, 0.2% gelatin.
Host Species Mouse
Immunogen full-length, recombinant, human TBK1 fused to a His·Tag™ sequence
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽80kDa TBK1 protein in HEK293 cells.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.