Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBK1 Rabbit anti-Human, Mouse, Canine, Chicken, Clone: AOW9, MilliporeSigma™

Catalog No. 04856MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-856-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-856-MI Supplier MilliporeSigma Supplier No. 04856
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Detect TBK1 using this Anti-TBK1 Antibody, clone AOW9 validated for use in western blotting and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBK1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone AOW9
Conjugate Unconjugated
Formulation Cultured supernatant containing 0.05% sodium azide.
Gene Accession No. NP_037386
Host Species Rabbit
Immunogen Peptide corresponding to the sequence C...EEVS... (amino acids 642-645) of human TANK-binding kinase 1 (TBK1) with an N-terminal cysteine added for conjugation purposes.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) EC 2.7.11.1, FLJ11330, NAK, T2K
Target Species Canine, Chicken, Human, Mouse
Content And Storage −20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.