Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tbr1 Chicken anti-Human, Mouse, Rat, Horse, Pig, Opossum, Bovine, Canine, Polyclonal, MilliporeSigma™

Catalog No. AB2261MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2261MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2261MI Supplier MilliporeSigma Supplier No. AB2261
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

Anti-Tbr1 Antibody is an antibody against Tbr1 for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tbr1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified chicken polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Symbols Tbr1; T-brain-1; TBR-1; TES-56
Host Species Chicken
Immunogen KLH-conjugated linear peptide corresponding to mouse Tbr1.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_033348
Target Species Bovine, Canine, Equine, Human, Mouse, Opossum, Pig, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.