Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tbr2 Rabbit anti-Mouse, Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB2283
Description
This Anti-Tbr2 Antibody is validated for use in immunohistochemistry for the detection of Tbr2.
Specifications
| Tbr2 | |
| Polyclonal | |
| Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Human, Mouse |
| Immunohistochemistry | |
| Unconjugated | |
| eomes; TBR2; eomesodermin; tbr1 | |
| KLH-conjugated linear peptide corresponding to mouse Tbr2. | |
| 100 μg | |
| NP_034266 | |
| 2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction