Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tbx1 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB15624MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB15624MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB15624MI Supplier MilliporeSigma Supplier No. AB15624
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Tbx1 Antibody is a Rabbit Polyclonal Antibody for detection of Tbx1 also known as T-box 1 and has been validated in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tbx1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid in PBS with 0.02% sodium azide.
Gene Symbols TBX1; CTHM; VCFS; brachyury; TGA; TBX1C; DGS; DORV; CAFS; DGCR
Host Species Rabbit
Immunogen Synthetic peptide from human Tbx1.
Purification Method Immunoaffinity purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_005992.1; NM_080646.1; NM_080647.1
Target Species Human
Content And Storage −20°C for 6 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.