Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

T-box 18, Mouse, Clone: 3C12, Abnova™

Catalog No. 89001113 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-113 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-113 Supplier Abnova Corporation Supplier No. H00009096M04
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant TBX18.

Sequence: STLLQGTGNGVPATHPHLLSGSSCSSPAFHLGPNTSQLCSLAPADYSACARSGLTLNRYSTSLAETYNRLTNQAGETFAPPRTPSYVGVSSSTSVNMSMGGTDGDTF

Specifications

Antigen T-box 18
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3C12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant TBX18.
Formulation PBS with no preservative; pH 7.4
Gene TBX18
Gene Accession No. XM_496819
Gene Symbols TBX18
Host Species Mouse
Immunogen TBX18 (XP_496819, 454 a.a. ∼ 560 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 9096
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.