Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TMEM229A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524405
Description

Specifications
TMEM229A | |
Polyclonal | |
TMEM229A | |
tcag7.977 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
319832, 730130 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
B2RXF0, B9EJI9 | |
TMEM229A | |
KLH conjugated synthetic peptide between 158-186 amino acids from the central region of human TMEM229A | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction